Learn More
Fine Science Tools Metzenbaum Baby Scissors, Stainless Steel, 14.5 cm
SDP
Catalog No. NC1549196
Encompass
Tips:Blunt-Blunt, Alloy / Material:Stainless Steel,Cutting Edge:35mm,Length:14.5cm,Tip Shape:Curved
This item is not returnable. View return policy
Catalog No. NC1549196 Supplier Fine Science Tools Supplier No. 1401914
Tips:Blunt-Blunt, Alloy / Material:Stainless Steel,Cutting Edge:35mm,Length:14.5cm,Tip Shape:Curved