Learn More
RWD LIFE SCIENCE INC METZENBAUM DISSECTING SCISSORS
SDP
Catalog No. NC2946614
Encompass
Straight/14.5cm; Blunt&Blunt; Cutting Edge(Length):40mm Width:6.3mm
Catalog No. NC2946614 Supplier RWD LIFE SCIENCE INC Supplier No. S1300514
Straight/14.5cm; Blunt&Blunt; Cutting Edge(Length):40mm Width:6.3mm