Learn More
Sklar Instruments Merit Metzenbaum-Lahey Dissecting Scissors, 5-3/4 in., Blunt/Blunt, Curved, Smooth, Stainless Steel, EA.
SDP
Catalog No. 501109769 Shop All Sklar Instruments Products
Encompass_Preferred
Merit Metzenbaum-Lahey Dissecting Scissors, 5-3/4 in., Blunt/Blunt, Curved, Smooth, Stainless Steel, EA. Merit Metzenbaum-Lahey Dissecting Scissors are most commonly used for dissecting and cutting tissue. These scissors are not recommended for cutting sutures, drains, or heavy tissue. Metzenbaum-Lahey scissors tend to be shorter than traditional Metzenbaum scissors. Metzenbaum Scissors are identifiable by their long, thin shanks and jaws that are relatively short. They also have fine, blunt, rounded tips. Merit instruments are Sklars office-grade line of products and most commonly utilized for routine and in-office minor procedures. This product has curved, smooth blades, blunt tips, and an overall length of 5-3/4 inches.
Catalog No. 50-110-9769 Supplier Sklar Instruments Supplier No. 97336
Merit Metzenbaum-Lahey Dissecting Scissors, 5-3/4 in., Blunt/Blunt, Curved, Smooth, Stainless Steel, EA. Merit Metzenbaum-Lahey Dissecting Scissors are most commonly used for dissecting and cutting tissue. These scissors are not recommended for cutting sutures, drains, or heavy tissue. Metzenbaum-Lahey scissors tend to be shorter than traditional Metzenbaum scissors. Metzenbaum Scissors are identifiable by their long, thin shanks and jaws that are relatively short. They also have fine, blunt, rounded tips. Merit instruments are Sklars office-grade line of products and most commonly utilized for routine and in-office minor procedures. This product has curved, smooth blades, blunt tips, and an overall length of 5-3/4 inches.