Learn More
Roboz Surgical METZENBAUM LAHEY SCS 7 IN CR
SDP
Catalog No. 5000574855
Encompass
These 7-inch Metzenbaum (Lahey) surgical scissors are designed for extra delicate dissection, featuring a curved profile and specialized construction for precise control during surgical procedures. The instrument is engineered with tungsten carbide reinforced blades, providing enhanced cutting performance and extended service life compared to traditional stainless steel alternatives.
  • Features tungsten carbide reinforced blades for superior cutting edge retention.
  • Designed with a curved configuration for improved visibility and access in delicate tissue applications.
  • Provides a 35mm blade length for precise, controlled cutting.
  • Offers increased durability with a lifespan over four times longer than standard stainless steel instruments.
Catalog No. 50-005-74855 Supplier Roboz Surgical Supplier No. RS6955SC
These 7-inch Metzenbaum (Lahey) surgical scissors are designed for extra delicate dissection, featuring a curved profile and specialized construction for precise control during surgical procedures. The instrument is engineered with tungsten carbide reinforced blades, providing enhanced cutting performance and extended service life compared to traditional stainless steel alternatives.
  • Features tungsten carbide reinforced blades for superior cutting edge retention.
  • Designed with a curved configuration for improved visibility and access in delicate tissue applications.
  • Provides a 35mm blade length for precise, controlled cutting.
  • Offers increased durability with a lifespan over four times longer than standard stainless steel instruments.