Learn More
Roboz Surgical METZENBAUM LAHEY SCS 7IN CRV
SDP
Catalog No. 5000574853
Encompass
These Metzenbaum (Lahey) scissors are designed for delicate surgical dissection and cutting. Featuring a curved geometry and 35mm blade length, this 7 in. instrument offers enhanced precision and accessibility during surgical procedures.
  • Curved blade profile for improved access to deep tissue
  • Overall length of 7 in. for optimal leverage and control
  • Precision-engineered 35mm blade for clean dissection
  • Designed for durability in demanding surgical environments
Catalog No. 50-005-74853 Supplier Roboz Surgical Supplier No. RS6953
These Metzenbaum (Lahey) scissors are designed for delicate surgical dissection and cutting. Featuring a curved geometry and 35mm blade length, this 7 in. instrument offers enhanced precision and accessibility during surgical procedures.
  • Curved blade profile for improved access to deep tissue
  • Overall length of 7 in. for optimal leverage and control
  • Precision-engineered 35mm blade for clean dissection
  • Designed for durability in demanding surgical environments