Learn More
Roboz Surgical METZENBAUM NEEDLE HOLDER 7IN S
SDP
Catalog No. 5000574566
Encompass
This Metzenbaum-style needle holder is a 7-inch surgical instrument designed for precision suturing in delicate procedural applications. Featuring a straight configuration, it is engineered for consistent control and durability.
  • Straight design facilitates precise needle positioning
  • Seven-inch length provides optimal reach and maneuverability
  • Integrated serrated jaws ensure a secure, non-slip grip on needles
Catalog No. 50-005-74566 Supplier Roboz Surgical Supplier No. RS7900
This Metzenbaum-style needle holder is a 7-inch surgical instrument designed for precision suturing in delicate procedural applications. Featuring a straight configuration, it is engineered for consistent control and durability.
  • Straight design facilitates precise needle positioning
  • Seven-inch length provides optimal reach and maneuverability
  • Integrated serrated jaws ensure a secure, non-slip grip on needles