Learn More
Roboz Surgical METZENBAUM SCISSOR CURVED 27MM
SDP
Catalog No. NC3585371
Encompass
These Metzenbaum scissors are designed for delicate surgical dissection and tissue manipulation. The instrument features a curved profile with blunt tips and a 27 mm blade length, providing precise control during fine procedures. With an overall length of 5 in. (127 mm), these scissors are suitable for detailed laboratory research applications.
  • Curved geometry for enhanced visibility in confined surgical sites
  • Blunt tips to minimize tissue trauma during blunt dissection
  • Overall length of 5 in. (127 mm)
  • Designed for laboratory and research use
  • Guaranteed free from defects in workmanship and material
Catalog No. NC3585371 Supplier Roboz Surgical Supplier No. RS6013
These Metzenbaum scissors are designed for delicate surgical dissection and tissue manipulation. The instrument features a curved profile with blunt tips and a 27 mm blade length, providing precise control during fine procedures. With an overall length of 5 in. (127 mm), these scissors are suitable for detailed laboratory research applications.
  • Curved geometry for enhanced visibility in confined surgical sites
  • Blunt tips to minimize tissue trauma during blunt dissection
  • Overall length of 5 in. (127 mm)
  • Designed for laboratory and research use
  • Guaranteed free from defects in workmanship and material