Learn More
World Precision Instrument Metzenbaum Scissors, 11.5cm, SuperCut, Curved
SDP
Catalog No. 502539515
Encompass
The shaft of Metzenbaum scissors is longer than the blades. They are used to cut delicate tissue or dissect blunt tissue. The tips are semi-blunt, and they are available in curved or straight styles.
Catalog No. 50-253-9515 Supplier World Precision Instrument Supplier No. 503270
The shaft of Metzenbaum scissors is longer than the blades. They are used to cut delicate tissue or dissect blunt tissue. The tips are semi-blunt, and they are available in curved or straight styles.