Learn More
World Precision Instrument Metzenbaum Scissors, 14cm, Straight, Titanium
SDP
Catalog No. 502539484
Encompass
The shaft of Metzenbaum scissors is longer than the blades. They are used to cut delicate tissue or dissect blunt tissue. The tips are semi-blunt, and they are available in curved or straight styles.
Catalog No. 50-253-9484 Supplier World Precision Instrument Supplier No. 503382
The shaft of Metzenbaum scissors is longer than the blades. They are used to cut delicate tissue or dissect blunt tissue. The tips are semi-blunt, and they are available in curved or straight styles.