Learn More
Roboz Surgical METZENBAUM SCISSORS 5 STRAIGH
SDP
Catalog No. NC0469599
Encompass
Metzenbaum Scissors; Sure Cut; Straight; Blunt; 25mm Blade Length; 5 in. Overall Length
Catalog No. NC0469599 Supplier Roboz Surgical Supplier No. RS6010SC
Metzenbaum Scissors; Sure Cut; Straight; Blunt; 25mm Blade Length; 5 in. Overall Length