Learn More
Electron Microscopy Sciences EMS SuperCut Metzenbaum Scissors Standard Curved, 9"
SDP
Catalog No. 501931413
Encompass_Preferred
Metzenbaum surgical scissors have rounded tips. SuperCut scissors are made in Germany using the highest grade of surgical steel and are tempered and hardened for an extended and sharper life than ordinary steel. Rounded and polished finger rings Curved version cuts into heavier or deeper tissue.
Blade: Standard Curved
OAL: 9"
This item is not returnable. View return policy
Catalog No. 50-193-1413 Supplier Electron Microscopy Sciences Supplier No. 7299606
Metzenbaum surgical scissors have rounded tips. SuperCut scissors are made in Germany using the highest grade of surgical steel and are tempered and hardened for an extended and sharper life than ordinary steel. Rounded and polished finger rings Curved version cuts into heavier or deeper tissue.
Blade: Standard Curved
OAL: 9"