Learn More
Braintree Scientific Metzenbaum Scissors Curved,Std Pattern 5.5""""
SDP
Catalog No. 501955874
Encompass
Metzenbaum Scissors Curved - Offering a unique combination of narrow blunted points along with thin delicate shafts for delicate dissection and minimal damage to surrounding tissues. **Sold for research purposes only and not for human use**
Catalog No. 50-195-5874 Supplier Braintree Scientific Supplier No. SCM180
Metzenbaum Scissors Curved - Offering a unique combination of narrow blunted points along with thin delicate shafts for delicate dissection and minimal damage to surrounding tissues. **Sold for research purposes only and not for human use**