Learn More
World Precision Instrument Metzenbaum Scissors Full Curve 14 cm
SDP
Catalog No. 502538310
Encompass
The shaft of Metzenbaum scissors is longer than the blades. They are used to cut delicate tissue or dissect blunt tissue. The tips are semi-blunt, and they are available in curved or straight styles.
Catalog No. 50-253-8310 Supplier World Precision Instrument Supplier No. WP4099
The shaft of Metzenbaum scissors is longer than the blades. They are used to cut delicate tissue or dissect blunt tissue. The tips are semi-blunt, and they are available in curved or straight styles.