Learn More
Roboz Surgical METZENBAUM SCS 5IN CV B/B SCUT
SDP
Catalog No. NC9400458
Encompass
These 5-inch curved Metzenbaum scissors feature advanced Sure Cut technology designed for high-precision tissue dissection. The instrument utilizes one micro-serrated blade to grip delicate or tough tissues, while the opposing blade is bevel-honed to a razor-sharp edge to ensure smooth, clean cuts without crushing. Identified by matte grey rings and a black screw, these scissors are engineered for effortless operation with reduced manual force.
  • Micro-serrated blade edge prevents tissue slippage
  • Razor-honed blade provides smooth, clean cutting performance
  • Matte grey handle rings and black screw for easy identification
  • Curved design with blunt tips for controlled dissection
  • Reduced force requirement for improved user efficiency
  • Designed exclusively for soft tissue applications
Catalog No. NC9400458 Supplier Roboz Surgical Supplier No. RS6013SC
These 5-inch curved Metzenbaum scissors feature advanced Sure Cut technology designed for high-precision tissue dissection. The instrument utilizes one micro-serrated blade to grip delicate or tough tissues, while the opposing blade is bevel-honed to a razor-sharp edge to ensure smooth, clean cuts without crushing. Identified by matte grey rings and a black screw, these scissors are engineered for effortless operation with reduced manual force.
  • Micro-serrated blade edge prevents tissue slippage
  • Razor-honed blade provides smooth, clean cutting performance
  • Matte grey handle rings and black screw for easy identification
  • Curved design with blunt tips for controlled dissection
  • Reduced force requirement for improved user efficiency
  • Designed exclusively for soft tissue applications