Learn More
Roboz Surgical METZENBAUM SCS 7IN CRV TC X-DE
SDP
Catalog No. 5000574407
Encompass
These Metzenbaum scissors are designed for fine surgical dissection, featuring a 7-inch curved profile tailored for delicate tissue work. The construction incorporates high-quality tungsten carbide inserts, providing enhanced edge retention and durability in demanding surgical environments.
  • Curved geometry facilitates precise cutting in confined spaces
  • Extra delicate design allows for refined manipulation of soft tissue
  • Tungsten carbide edges ensure long-lasting sharpness and reliable performance
  • Crafted for professional surgical and laboratory applications
Catalog No. 50-005-74407 Supplier Roboz Surgical Supplier No. RS6956
These Metzenbaum scissors are designed for fine surgical dissection, featuring a 7-inch curved profile tailored for delicate tissue work. The construction incorporates high-quality tungsten carbide inserts, providing enhanced edge retention and durability in demanding surgical environments.
  • Curved geometry facilitates precise cutting in confined spaces
  • Extra delicate design allows for refined manipulation of soft tissue
  • Tungsten carbide edges ensure long-lasting sharpness and reliable performance
  • Crafted for professional surgical and laboratory applications