Learn More
Roboz Surgical Roboz Metzenbaum Scissors, Straight, 7 in, SureCut, 1 ea
SDP
Catalog No. 5000574874
Encompass
These straight Metzenbaum scissors are precision surgical instruments engineered for delicate tissue dissection. Featuring the specialized SureCut technology, they provide clean, effortless cutting performance through a refined edge design. The instruments are identified by matte grey finger rings and a black screw for easy recognition.
  • Designed for delicate tissue dissection
  • Feature a razor edge blade for high-precision cutting
  • Constructed with a 7-inch overall length
  • Equipped with matte grey rings and a black screw for identification
  • Not intended for cutting bone, wire, or other hard materials which may damage the blades
Catalog No. 50-005-74874 Supplier Roboz Surgical Supplier No. RS6952SC
These straight Metzenbaum scissors are precision surgical instruments engineered for delicate tissue dissection. Featuring the specialized SureCut technology, they provide clean, effortless cutting performance through a refined edge design. The instruments are identified by matte grey finger rings and a black screw for easy recognition.
  • Designed for delicate tissue dissection
  • Feature a razor edge blade for high-precision cutting
  • Constructed with a 7-inch overall length
  • Equipped with matte grey rings and a black screw for identification
  • Not intended for cutting bone, wire, or other hard materials which may damage the blades