Learn More
Electron Microscopy Sciences EMS SuperCut Metzenbaum Scissors Delicate Curved Blades, 5"
SDP
Catalog No. 501931411
Encompass_Preferred
Metzenbaum surgical scissors have rounded tips. SuperCut scissors are made in Germany using the highest grade of surgical steel and are tempered and hardened for an extended and sharper life than ordinary steel. Rounded and polished finger rings Curved version cuts into heavier or deeper tissue.
Blade: Delicate Curved
OAL: 5"
This item is not returnable. View return policy
Catalog No. 50-193-1411 Supplier Electron Microscopy Sciences Supplier No. 7299604
Metzenbaum surgical scissors have rounded tips. SuperCut scissors are made in Germany using the highest grade of surgical steel and are tempered and hardened for an extended and sharper life than ordinary steel. Rounded and polished finger rings Curved version cuts into heavier or deeper tissue.
Blade: Delicate Curved
OAL: 5"