Learn More
Electron Microscopy Sciences EMS SuperCut Metzenbaum Scissors with Delicate Straight Blades, 5"
SDP
Catalog No. 501931410
Encompass_Preferred
Metzenbaum surgical scissors have rounded tips. SuperCut scissors are made in Germany using the highest grade of surgical steel and are tempered and hardened for an extended and sharper life than ordinary steel. Rounded and polished finger rings Straight version cuts into surface area tissue.
Blade: Delicate Straight
OAL: 5"
This item is not returnable. View return policy
Catalog No. 50-193-1410 Supplier Electron Microscopy Sciences Supplier No. 7299603
Metzenbaum surgical scissors have rounded tips. SuperCut scissors are made in Germany using the highest grade of surgical steel and are tempered and hardened for an extended and sharper life than ordinary steel. Rounded and polished finger rings Straight version cuts into surface area tissue.
Blade: Delicate Straight
OAL: 5"