Learn More
Medchemexpress LLC Mevalonate 5-phosphate trilithium | 868553-49-5 | 99.0% | 245.94 | 1 MG
SDP
Catalog No. 5000393898
Encompass_Preferred
Mevalonate 5-phosphate trilithium is a substrate of phosphomevalonate kinase and mevalonate-5-phosphate decarboxylase. It is for research use only and not sold to patients.
Catalog No. 50-003-93898 Supplier Medchemexpress LLC Supplier No. HY164091A1MG
Mevalonate 5-phosphate trilithium is a substrate of phosphomevalonate kinase and mevalonate-5-phosphate decarboxylase. It is for research use only and not sold to patients.