Learn More
Medchemexpress LLC Mevalonate kinase (MVK), human (Sf9, His-GST) | >85.0% | 10 UG
SDP
Catalog No. 5000294268
Encompass_Preferred
Recombinant full-length human mevalonate kinase (MVK) produced in Sf9 insect cells and supplied as an N-terminal His- and GST-tagged protein in buffered solution. Designed for research use in enzymatic assays, structural studies, and antibody generation. Supplied as a filtered solution formulated for stability and low endotoxin, and shipped on dry ice for cryogenic storage.
Catalog No. 50-002-94268 Supplier Medchemexpress LLC Supplier No. HYP7473910UG
Recombinant full-length human mevalonate kinase (MVK) produced in Sf9 insect cells and supplied as an N-terminal His- and GST-tagged protein in buffered solution. Designed for research use in enzymatic assays, structural studies, and antibody generation. Supplied as a filtered solution formulated for stability and low endotoxin, and shipped on dry ice for cryogenic storage.