Learn More
Medchemexpress LLC HY-113071A 50mg , Mevalonic acid (lithium salt) CAS:2618458-93-6 Purity:>98%
SDP
Catalog No. 502260558
Encompass_Preferred
HY-113071A 50mg Mevalonic acid (lithium salt) CAS: 2618458-93-6 Mevalonic acid lithium salt, a precursor in the mevalonate pathway, is essential for cell growth and proliferation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
This item is not returnable. View return policy
Catalog No. 50-226-0558 Supplier Medchemexpress LLC Supplier No. HY113071A50MG
HY-113071A 50mg Mevalonic acid (lithium salt) CAS: 2618458-93-6 Mevalonic acid lithium salt, a precursor in the mevalonate pathway, is essential for cell growth and proliferation. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.