Learn More
Medchemexpress LLC Mevalonic acid lithium salt | 2618458-93-6 | 98.0% | 154.09 | 250 MG
SDP
Catalog No. 5000439909
Encompass_Preferred
Mevalonic acid lithium salt, also known as MVA lithium salt, is a precursor in the mevalonate pathway, which plays a vital role in cell growth and proliferation. It has been shown to inhibit simvastatin-induced decreases in C2C12 cell viability in vitro and is relevant for research into myopathy and heart failure.
  • Essential for cell growth and proliferation studies.
  • Inhibits simvastatin-induced decrease in C2C12 cell viability.
  • Useful for myopathy and heart failure research.
  • Shortens activated partial thromboplastin time (APTT) and prothrombin time (PT) in mice.
  • Increases thrombus weight in mice.
  • Increases infarct volume after transient middle cerebral artery occlusion in mice.
  • Determines cell proliferation in HCT116 and LoVo cells.
  • Assesses changes in YAP expression in HOS and MG63 cells.
Catalog No. 50-004-39909 Supplier Medchemexpress LLC Supplier No. 250MGHY113071A
Mevalonic acid lithium salt, also known as MVA lithium salt, is a precursor in the mevalonate pathway, which plays a vital role in cell growth and proliferation. It has been shown to inhibit simvastatin-induced decreases in C2C12 cell viability in vitro and is relevant for research into myopathy and heart failure.
  • Essential for cell growth and proliferation studies.
  • Inhibits simvastatin-induced decrease in C2C12 cell viability.
  • Useful for myopathy and heart failure research.
  • Shortens activated partial thromboplastin time (APTT) and prothrombin time (PT) in mice.
  • Increases thrombus weight in mice.
  • Increases infarct volume after transient middle cerebral artery occlusion in mice.
  • Determines cell proliferation in HCT116 and LoVo cells.
  • Assesses changes in YAP expression in HOS and MG63 cells.