Learn More
Medchemexpress LLC Mevalonolactone | 19115-49-2 | MFCD01074894 | 98.0% | 130.14 g/mol | C6H10O3 | 5 MG

Supplier: Medchemexpress LLC HY1357005MG
Mevalonolactone is a sesquiterpene lactone used in laboratory research as a calcineurin inhibitor. It has been reported to inhibit calcineurin in a p-Nitrophenyl phosphate assay with an IC50 of 134.29 μM. The material is supplied as a high-purity research chemical in small, mg-scale quantities for biochemical and cell-based studies.
- Displays calcineurin inhibitory activity (IC50 134.29 μM).
- High purity: 98.0%.
- Molecular weight: 130.14 g/mol.
- Chemical formula: C6H10O3.
- Supplied in multiple research-scale package sizes.
- Storage guidance for solvent solutions: -80°C (2 years) or -20°C (1 year).
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.