Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

(±)-Mevalonolactone, 97%

Catalog No. p-4089510
Change view
Click to view available options
Quantity:
1 g
5 g
Packaging:
Glass Bottle
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC428300010 1 g Glass Bottle
AC428300050 5 g Glass Bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. AC428300010 Supplier Thermo Scientific Chemicals Supplier No. 428300010
Only null left
Add to Cart
This item is not returnable. View return policy

CAS: 674-26-0 | C6H10O3 | 130.14 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 674-26-0
Molecular Formula C6H10O3
Molecular Weight (g/mol) 130.14
MDL Number MFCD00006648
InChI Key JYVXNLLUYHCIIH-UHFFFAOYNA-N
Synonym mevalonolactone, dl-mevalonic acid lactone, dl-mevalonolactone, 4-hydroxy-4-methyltetrahydro-2h-pyran-2-one, mevalonic acid lactone, mevalonic lactone, dl-mevalolactone, 2h-pyran-2-one, tetrahydro-4-hydroxy-4-methyl, 2h-pyran-2-one, tetrahydro-4-hydroxy-4-methyl-, .+/-., 1-tetrahydro-4-hydroxy-4-methyl-2h-pyran-2-one
PubChem CID 10428
IUPAC Name 4-hydroxy-4-methyloxan-2-one
SMILES CC1(O)CCOC(=O)C1

Specifications

CAS Min % 96.0
CAS Max % 100.0
Melting Point 28.0°C
Boiling Point 145.0°C to 150.0°C (5.0 mmHg)
Flash Point >110°C
Assay Percent Range 96% min. (GC)
Packaging Glass Bottle
Quantity 1 g
Solubility Information Solubility in water: soluble
Formula Weight 130.14
Percent Purity 97%
Chemical Name or Material (±)-Mevalonolactone
Show More Show Less
EINECSNumber 211-615-

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.