Learn More
Selleck Chemical LLC Mevastatin 50mg 73573-88-3 ML-236B
SDP
Supplier:  Selleck Chemical LLC S422350MG
Encompass_Preferred
Mevastatin (ML-236B) is a competitive inhibitor of HMG-Coenzyme A (HMG-CoA) reductase with a binding affinity 10,000 times greater than the HMG-CoA substrate itself. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. NC1085598