Learn More
Medchemexpress LLC Mevastatin | 73573-88-3 | 99.46% | 390.51 | 100 MG
SDP
Catalog No. 5000385757
Encompass_Preferred
Mevastatin (Compactin) is a first HMG-CoA reductase inhibitor belonging to the statin class. It functions as a lipid-lowering agent and is known to induce apoptosis and arrest cancer cells in the G0/G1 phase. Additionally, Mevastatin increases endothelial nitric oxide synthase (eNOS) mRNA and protein levels. It exhibits antitumor activity and shows potential for the treatment of cardiovascular diseases.
  • Primary inhibitor of HMG-CoA reductase
  • Lipid-lowering agent
  • Induces apoptosis
  • Arrests cancer cells in the G0/G1 phase
  • Increases eNOS mRNA and protein levels
  • Exhibits antitumor activity
  • Potential for cardiovascular disease treatment
  • Initial source from Microorganisms (Penicillium citrinum)
  • Solid, white to off-white appearance
Catalog No. 50-003-85757 Supplier Medchemexpress LLC Supplier No. HY17408100MG
Mevastatin (Compactin) is a first HMG-CoA reductase inhibitor belonging to the statin class. It functions as a lipid-lowering agent and is known to induce apoptosis and arrest cancer cells in the G0/G1 phase. Additionally, Mevastatin increases endothelial nitric oxide synthase (eNOS) mRNA and protein levels. It exhibits antitumor activity and shows potential for the treatment of cardiovascular diseases.
  • Primary inhibitor of HMG-CoA reductase
  • Lipid-lowering agent
  • Induces apoptosis
  • Arrests cancer cells in the G0/G1 phase
  • Increases eNOS mRNA and protein levels
  • Exhibits antitumor activity
  • Potential for cardiovascular disease treatment
  • Initial source from Microorganisms (Penicillium citrinum)
  • Solid, white to off-white appearance