Learn More
Enzo Life Sciences Mevastatin sodium (5mg). CAS: 99782-89-5
SDP
Supplier:  Enzo Life Sciences BMLG2380005
Encompass_Preferred
This is the active carboxylate form of mevastatin. It is active in whole cells as well as in cell free assays. Purity: ≥98% (HPLC). Solubility: Soluble in ethanol (20 mg/ml) and DMSO (30 mg/ml). Long Term Storage: -20°C.
Catalog No. 50-201-0871