Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mevastatin, Tocris Bioscience™

Catalog No. 152610
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
15-261-0 10 mg
15-265-0 50 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 15-261-0 Supplier R&D Systems Supplier No. 1526/10

Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.

This item is not returnable. View return policy

HMG-CoA reductase inhibitor

Inhibitor of HMG-CoA reductase; decreases cholesterol biosynthesis, in vitro and in vivo. Induces apoptosis, arrests cancer cells in G1 phase and downregulates cdk 2, 4, and 6, cyclin D1 and E1, p21 and p27. Inactive lactam prodrug of mevastatin hydroxy acid, naturally bioactivated in vivo following oral administration.

Chemical Identifiers

CAS 73573-88-3
Molecular Formula C23H34O5
Molecular Weight (g/mol) 390.52
MDL Number MFCD05662341
InChI Key AJLFOPYRIVGYMJ-INTXDZFKSA-N
Synonym mevastatin, compactin, mevastatinum, mevastatina, mevastatine, mevastatinum inn-latin, antibiotic ml 236b, mevastatin inn, compactin penicillium, mevastatine inn-french
PubChem CID 64715
ChEBI CHEBI:34848
IUPAC Name (1S,7S,8S,8aR)-8-{2-[(2R,4R)-4-hydroxy-6-oxooxan-2-yl]ethyl}-7-methyl-1,2,3,7,8,8a-hexahydronaphthalen-1-yl (2S)-2-methylbutanoate
SMILES CC[C@H](C)C(=O)O[C@H]1CCC=C2C=C[C@H](C)[C@H](CC[C@@H]3C[C@@H](O)CC(=O)O3)[C@@H]12

Specifications

Quantity 10 mg
Formula Weight 390.52
Percent Purity >98%
Chemical Name or Material Mevastatin
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.