Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Mevastatin, Tocris Bioscience™
Description
Inhibitor of HMG-CoA reductase; decreases cholesterol biosynthesis, in vitro and in vivo. Induces apoptosis, arrests cancer cells in G1 phase and downregulates cdk 2, 4, and 6, cyclin D1 and E1, p21 and p27. Inactive lactam prodrug of mevastatin hydroxy acid, naturally bioactivated in vivo following oral administration.
Specifications
Specifications
| Quantity | 50 mg |
| Formula Weight | 390.52 |
| Percent Purity | >98% |
| Chemical Name or Material | Mevastatin |
Safety and Handling
| Recommended Storage | Store at Room Temperature |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?