Learn More
ATCC MeWo; Melanoma; Human (Homo sapiens)
Catalog No. 502382194
Encompass
MeWo is a cell line exhibiting fibroblast morphology that was isolated from the skin of a 78-year-old, White male patient with malignant melanoma. This product has applications in toxicology.
Catalog No. 50-238-2194 Supplier ATCC Supplier No. HTB65
MeWo is a cell line exhibiting fibroblast morphology that was isolated from the skin of a 78-year-old, White male patient with malignant melanoma. This product has applications in toxicology.