Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Medchemexpress LLC Mexiletine | 31828-71-4 | 99.9% | 250 MG
SDP

Catalog No. 5000231808
Encompass_Preferred

Mexiletine is a Class IB antiarrhythmic agent and sodium-channel blocker supplied as a research-grade chemical for laboratory use. It is used in cardiovascular and neurological research to study sodium-channel inhibition and antiarrhythmic effects; not for human therapeutic use.

  • Class IB antiarrhythmic and sodium-channel blocker.
  • Research-grade chemical for laboratory use.
  • Purity 99.9% (as supplied).
  • Molecular formula C11H17NO; molecular weight 179.26 g/mol.
  • Available as a 250 mg pack suitable for small-scale studies.

Catalog No. 50-002-31808 Supplier Medchemexpress LLC Supplier No. HY119521250MG
May include imposed supplier surcharges.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Mexiletine is a Class IB antiarrhythmic agent and sodium-channel blocker supplied as a research-grade chemical for laboratory use. It is used in cardiovascular and neurological research to study sodium-channel inhibition and antiarrhythmic effects; not for human therapeutic use.

  • Class IB antiarrhythmic and sodium-channel blocker.
  • Research-grade chemical for laboratory use.
  • Purity 99.9% (as supplied).
  • Molecular formula C11H17NO; molecular weight 179.26 g/mol.
  • Available as a 250 mg pack suitable for small-scale studies.

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.