Learn More
Medchemexpress LLC Mexiletine-d6 hydrochloride | 1329835-60-0 | 99.9% | 221.76 | 5 MG
SDP
Catalog No. 5000393453
Encompass_Preferred
Mexiletine-d6 hydrochloride is a deuterium-labeled compound, serving as a tracer and internal standard for quantitative analysis methods such as NMR, GC-MS, or LC-MS. It is a Class IB antiarrhythmic and a non-selective voltage-gated sodium channel blocker.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis
  • Functions as a class IB antiarrhythmic
  • Acts as a non-selective voltage-gated sodium channel blocker
  • Suitable for research use only
Catalog No. 50-003-93453 Supplier Medchemexpress LLC Supplier No. 5MGHYA0093S
Mexiletine-d6 hydrochloride is a deuterium-labeled compound, serving as a tracer and internal standard for quantitative analysis methods such as NMR, GC-MS, or LC-MS. It is a Class IB antiarrhythmic and a non-selective voltage-gated sodium channel blocker.
  • Can be used as a tracer
  • Can be used as an internal standard for quantitative analysis
  • Functions as a class IB antiarrhythmic
  • Acts as a non-selective voltage-gated sodium channel blocker
  • Suitable for research use only