Learn More
Medchemexpress LLC Mexiletine-d6 hydrochloride | 1329835-60-0 | 99.9% | C11H12D6ClNO | 1 MG
SDP
Catalog No. 5000432146
Encompass_Preferred
Mexiletine-d6 hydrochloride is a deuterium labeled Mexiletine hydrochloride. It acts as a Class IB antiarrhythmic and a non-selective voltage-gated sodium channel blocker.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.
Catalog No. 50-004-32146 Supplier Medchemexpress LLC Supplier No. HYA0093S1MG
Add to Cart
Add to Cart
Mexiletine-d6 hydrochloride is a deuterium labeled Mexiletine hydrochloride. It acts as a Class IB antiarrhythmic and a non-selective voltage-gated sodium channel blocker.
  • Can be used as a tracer.
  • Can be used as an internal standard for quantitative analysis by NMR, GC-MS, or LC-MS.