Learn More
Apexbio Technology LLC Mexiletine HCl 5370-01-4 10mM (in 1mL DMSO)
SDP
Catalog No. 5000094230
Encompass_Preferred
Mexiletine hydrochloride (CAS 5370-01-4) is a small molecule classified as a Class IB anti-arrhythmic agent It modulates cardiac electrophysiological activity primarily by inhibiting voltage-gated sodium channels resulting in decreased inward sodium currents during the cardiac action potential Through this mechanism mexiletine HCl stabilizes cardiac cell membranes and suppresses abnormal electrical activity This compound is frequently utilized in research to study sodium channel function cardiac arrhythmia models and the pharmacodynamics of anti-arrhythmic interventions
Catalog No. 50-000-94230 Supplier Apexbio Technology LLC Supplier No. B17895.1
Mexiletine hydrochloride (CAS 5370-01-4) is a small molecule classified as a Class IB anti-arrhythmic agent It modulates cardiac electrophysiological activity primarily by inhibiting voltage-gated sodium channels resulting in decreased inward sodium currents during the cardiac action potential Through this mechanism mexiletine HCl stabilizes cardiac cell membranes and suppresses abnormal electrical activity This compound is frequently utilized in research to study sodium channel function cardiac arrhythmia models and the pharmacodynamics of anti-arrhythmic interventions