Learn More
Medchemexpress LLC Mexiletine hydrochloride | 5370-01-4 | 99.88% | 215.72 | 500 MG
SDP
Catalog No. 5000365665
Encompass_Preferred
Mexiletine hydrochloride is an orally effective antiarrhythmic agent used for myotonia and neuropathic pain. It blocks sodium channels, making it suitable for cardiovascular and neurological research. This product is for research use only.
  • Orally effective antiarrhythmic agent
  • Effective for myotonia and neuropathic pain
  • Blocks sodium channels
  • Suitable for cardiovascular and neurological research
Catalog No. 50-003-65665 Supplier Medchemexpress LLC Supplier No. 500MGHYA0093
Mexiletine hydrochloride is an orally effective antiarrhythmic agent used for myotonia and neuropathic pain. It blocks sodium channels, making it suitable for cardiovascular and neurological research. This product is for research use only.
  • Orally effective antiarrhythmic agent
  • Effective for myotonia and neuropathic pain
  • Blocks sodium channels
  • Suitable for cardiovascular and neurological research