Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mexiletine hydrochloride

Catalog No. AC372910050
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
AC372910050 5 g
AC372910250 25 g
AC372911000 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC372910050 Supplier Thermo Scientific Chemicals Supplier No. 372910050
Get Up to $1,800 in Chemicals with Purchase
View all promotions
Add to Cart

CAS: 5370-01-4 | C11H17NO·HCl | 215.72 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 1-4-5370
Molecular Formula C11H17NO·HCl
Molecular Weight (g/mol) 215.72
InChI Key NFEIBWMZVIVJLQ-UHFFFAOYNA-N
Synonym mexiletine hydrochloride, mexitil, mexiletine hcl, mexiletene hydrochloride, 1-2,6-dimethylphenoxy-2-propanamine hydrochloride, 1-2,6-dimethylphenoxy propan-2-amine hydrochloride, 2-propanamine, 1-2,6-dimethylphenoxy-, hydrochloride, mexitil tn, 1-2,6-xylyloxy-2-aminopropane hydrochloride
PubChem CID 21467
ChEBI CHEBI:6917
IUPAC Name 1-(2,6-dimethylphenoxy)propan-2-amine;hydrochloride
SMILES CC1=C(C(=CC=C1)C)OCC(C)N.Cl

Specifications

Melting Point 202.0°C
Quantity 5 g
Infrared Spectrum Authentic
Linear Formula (CH3)2C6H3OCH2CH(NH2)CH3·HCl
Merck Index 15,6248
Solubility Information Solubility in water: soluble. Other solubilities: soluble in alcohols
Formula Weight 215.72
Percent Purity ≥97.5% (HPLC)
Chemical Name or Material Mexiletine hydrochloride
Hazard Category GHS Signal Word: Warning
Hazard Statement GHS H Statement
Harmful if swallowed.
Causes skin irritation.
Causes serious eye irritation.
Precautionary Statement GHS P Statement
IF SWALLOWED: rinse mouth.
Do NOT induce vomiting.
Call a POISON CENTER or doctor/physician if you feel unwell.
IF ON SKIN: Wash with plenty of soap and water.
If eye irritation persists: Get medical advice/attent
EINECSNumber 226-362-1

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.