Learn More
Medchemexpress LLC HY-N7689 1mg Medchemexpress, Mexoticin CAS:18196-00-4 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HYN76891MG
Encompass_Preferred
Medchemexpress, HY-N7689 1mg Mexoticin CAS:18196-00-4 Mexoticin is a naturally occurring coumarin that can be isolated from the leaves of Murraya omphalocarpa. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-3009