Learn More
MSC Meyer Gage Gage Pin, 0.3456 in, Class X, Plus Tolerance, 52100 Steel
Catalog No. 5000534835
Encompass
This precision gage pin, made from durable 52100 steel, features a 0.3456 in diameter and Class X plus tolerance. It is ideal for accurate inspection needs, maximizing GO/NOGO tolerance limits, and calibrating various measuring equipment such as micrometers and calipers. Each pin over 0.060 in is laser marked with its size, class, tolerance direction, and a serial number for complete traceability.
  • Made from 52100 steel for durability.
  • Features a precise class X tolerance of 0.00004 in.
  • Ensures high accuracy with a roundness limit of 0.00002 in.
  • Standard length of 2 in for class X gage pins.
  • Laser marked with size, class, and tolerance direction for traceability on pins over 0.060 in.
  • Ideal for various inspection applications.
  • Maximizes go/nogo tolerance limits.
  • Can be used to calibrate measuring equipment, micrometers, and calipers.
Catalog No. 50-005-34835 Supplier MSC Supplier No. 43706522
This precision gage pin, made from durable 52100 steel, features a 0.3456 in diameter and Class X plus tolerance. It is ideal for accurate inspection needs, maximizing GO/NOGO tolerance limits, and calibrating various measuring equipment such as micrometers and calipers. Each pin over 0.060 in is laser marked with its size, class, tolerance direction, and a serial number for complete traceability.
  • Made from 52100 steel for durability.
  • Features a precise class X tolerance of 0.00004 in.
  • Ensures high accuracy with a roundness limit of 0.00002 in.
  • Standard length of 2 in for class X gage pins.
  • Laser marked with size, class, and tolerance direction for traceability on pins over 0.060 in.
  • Ideal for various inspection applications.
  • Maximizes go/nogo tolerance limits.
  • Can be used to calibrate measuring equipment, micrometers, and calipers.