Learn More
MSC Meyer Gage Pin Gage, 0.502 inch, Class Z Plus, Steel
Catalog No. 5000529042
Encompass
This pin gage is designed for precise measurement applications, featuring a 0.502 inch diameter with a Class Z Plus tolerance. Constructed from 52100 steel, it offers durability and accuracy. Each gage is 2 inches in length and is engineered to meet strict quality standards for verifying hole sizes and depths.
  • Constructed from 52100 steel
  • 2 inch standard length
  • Features a Class Z tolerance of .0001 inch
  • Plus (GO) tolerance direction for verifying minimum hole sizes
  • Roundness limit of .00005 inch
  • Heat treated to a Rockwell C Hardness 60-62 for durability
  • Laser marked with size, tolerance direction, and serial numbers for traceability (for pins over 0.060 inch)
Catalog No. 50-005-29042 Supplier MSC Supplier No. 55557615
This pin gage is designed for precise measurement applications, featuring a 0.502 inch diameter with a Class Z Plus tolerance. Constructed from 52100 steel, it offers durability and accuracy. Each gage is 2 inches in length and is engineered to meet strict quality standards for verifying hole sizes and depths.
  • Constructed from 52100 steel
  • 2 inch standard length
  • Features a Class Z tolerance of .0001 inch
  • Plus (GO) tolerance direction for verifying minimum hole sizes
  • Roundness limit of .00005 inch
  • Heat treated to a Rockwell C Hardness 60-62 for durability
  • Laser marked with size, tolerance direction, and serial numbers for traceability (for pins over 0.060 inch)