Learn More
World Precision Instrument Meyhoefer Curette, 13cm, Size 2 (2.25mm Diameter)
SDP
Catalog No. 502539289
Encompass
The Meyhoefer curette has an ice cream scoop-shaped tip with semi-sharp edges designed for scraping. In ophthalmology, it may be used to scrape chalazion from the eyelids. It is perfect for incising cysts, large and small. This curette has a sturdy handle with a smooth surface that improves the grip. Its square shape give you a solid grip and helps alleviate finger strain.
Catalog No. 50-253-9289 Supplier World Precision Instrument Supplier No. 504769
The Meyhoefer curette has an ice cream scoop-shaped tip with semi-sharp edges designed for scraping. In ophthalmology, it may be used to scrape chalazion from the eyelids. It is perfect for incising cysts, large and small. This curette has a sturdy handle with a smooth surface that improves the grip. Its square shape give you a solid grip and helps alleviate finger strain.