Learn More
Apexbio Technology LLC Mezlocillin Sodium 42057-22-7 10mM (in 1mL DMSO)
SDP
Catalog No. 5000096494
Encompass_Preferred
Mezlocillin Sodium (CAS 42057-22-7) is a small-molecule inhibitor targeting bacterial penicillin-binding proteins (PBPs) It is designed to inhibit bacterial cell wall synthesis thereby disrupting peptidoglycan cross-linking and compromising bacterial cell wall integrity Mezlocillin Sodium exerts its biological activity primarily through irreversible inhibition of PBPs leading to bacterial lysis and death In laboratory studies Mezlocillin Sodium demonstrates antibacterial inhibition with minimal inhibitory concentration (MIC) and half-maximal inhibitory concentration (IC50) values determined in susceptibility assays Based on these pharmacological properties Mezlocillin Sodium holds research potential in studies of bacterial susceptibility resistance mechanisms and antibiotic pharmacodynamics
Catalog No. 50-000-96494 Supplier Apexbio Technology LLC Supplier No. B17905.1
Mezlocillin Sodium (CAS 42057-22-7) is a small-molecule inhibitor targeting bacterial penicillin-binding proteins (PBPs) It is designed to inhibit bacterial cell wall synthesis thereby disrupting peptidoglycan cross-linking and compromising bacterial cell wall integrity Mezlocillin Sodium exerts its biological activity primarily through irreversible inhibition of PBPs leading to bacterial lysis and death In laboratory studies Mezlocillin Sodium demonstrates antibacterial inhibition with minimal inhibitory concentration (MIC) and half-maximal inhibitory concentration (IC50) values determined in susceptibility assays Based on these pharmacological properties Mezlocillin Sodium holds research potential in studies of bacterial susceptibility resistance mechanisms and antibiotic pharmacodynamics