Learn More
Medchemexpress LLC Mezlocillin (sodium) | 42057-22-7 | 98.1% | 200 MG
SDP
Catalog No. 5000413401
Encompass_Preferred
Mezlocillin (BAY-f 1353) sodium is a β-lactam antibiotic, a semisynthetic and extended-spectrum antibiotic. Mezlocillin sodium is active against both gram-negative and gram-positive bacteria. Mezlocillin sodium can be used in bacterial infection research.
  • Semisynthetic and extended-spectrum antibiotic
  • Active against both gram-negative and gram-positive bacteria
  • Can be used in bacterial infection research
Catalog No. 50-004-13401 Supplier Medchemexpress LLC Supplier No. HYB1466200MG
Mezlocillin (BAY-f 1353) sodium is a β-lactam antibiotic, a semisynthetic and extended-spectrum antibiotic. Mezlocillin sodium is active against both gram-negative and gram-positive bacteria. Mezlocillin sodium can be used in bacterial infection research.
  • Semisynthetic and extended-spectrum antibiotic
  • Active against both gram-negative and gram-positive bacteria
  • Can be used in bacterial infection research