Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Mezlocillin Sodium, USP Grade, G-Biosciences
SCI_ED

Catalog No. S139784
Change view
Click to view available options
Quantity:
500 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
S139784 500 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. S139784 Supplier G-Biosciences Supplier No. RC1052
Add to Cart
Add to Cart

Mezlocillin sodium is an extended-spectrum Beta-lactam antibiotic of the penicillin family.

It targets gram-negative and gram-positive bacteria.

Chemical Identifiers

Molecular Weight (g/mol) 561.56 g/mol

Specifications

Quantity 500 mg
Percent Purity 98%
Grade USP
Physical Form Powder
Product Line Gibco
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.