Learn More
TARGETMOL CHEMICALS INC MF-094 10MG
SDP
Catalog No. 502878415
Encompass_Preferred
Also available in 1 mL, 1 mg, 5 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. MF-094 is a selective USP30 inhibitor with IC50 of 0.12 uM. MF-094 can increase protein ubiquitination and accelerate mitophagy. Purity 98.46%
Catalog No. 50-287-8415 Supplier TARGETMOL CHEMICALS INC Supplier No. T1202410MG
Also available in 1 mL, 1 mg, 5 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. MF-094 is a selective USP30 inhibitor with IC50 of 0.12 uM. MF-094 can increase protein ubiquitination and accelerate mitophagy. Purity 98.46%