Learn More
Medchemexpress LLC HY-112438 10mg Medchemexpress, MF-094 CAS:2241025-68-1 Purity:>98%
SDP
Catalog No. 501969093
Encompass_Preferred
Medchemexpress, HY-112438 10mg MF-094 CAS:2241025-68-1 MF-094 is a potent and selective USP30 inhibitor with an IC50 of 120 nM. MF-094 increases protein ubiquitination and accelerates mitophagy. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-196-9093 Supplier Medchemexpress LLC Supplier No. HY11243810MG
Medchemexpress, HY-112438 10mg MF-094 CAS:2241025-68-1 MF-094 is a potent and selective USP30 inhibitor with an IC50 of 120 nM. MF-094 increases protein ubiquitination and accelerates mitophagy. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.