Learn More
Medchemexpress LLC MF-094 | 2241025-68-1 | 99.8% | 535.70 | 1 ML
SDP
Catalog No. 5000438245
Encompass_Preferred
MF-094 is a potent and selective inhibitor of USP30 with an IC50 of 120 nM. This compound has been shown to increase protein ubiquitination and accelerate mitophagy.
  • Potent and selective USP30 inhibitor
  • IC50 of 120 nM for USP30
  • Increases protein ubiquitination
  • Accelerates mitophagy
  • Has <30% inhibitory activity for a panel of 22 USPs assays at 10 uM
Catalog No. 50-004-38245 Supplier Medchemexpress LLC Supplier No. HY11243810MM/1ML
MF-094 is a potent and selective inhibitor of USP30 with an IC50 of 120 nM. This compound has been shown to increase protein ubiquitination and accelerate mitophagy.
  • Potent and selective USP30 inhibitor
  • IC50 of 120 nM for USP30
  • Increases protein ubiquitination
  • Accelerates mitophagy
  • Has <30% inhibitory activity for a panel of 22 USPs assays at 10 uM