Learn More
Medchemexpress LLC MF-094 | 2241025-68-1 | 99.8% | 535.70 | 50 MG
SDP
Catalog No. 5000365716
Encompass_Preferred
MF-094 is a potent and selective USP30 inhibitor, effective at an IC50 of 120 nM. This compound has been shown to increase protein ubiquitination and accelerate mitophagy, making it suitable for biochemical research applications. It exhibits high selectivity, with less than 30% inhibitory activity against a panel of 22 other USPs at 10 μM.
  • Potent and selective USP30 inhibitor
  • Increases protein ubiquitination
  • Accelerates mitophagy
  • Minimal inhibitory activity against other USPs
  • Stable as powder at -20°C for 3 years
  • Stable in solvent at -80°C for 2 years
Catalog No. 50-003-65716 Supplier Medchemexpress LLC Supplier No. 50MGHY112438
MF-094 is a potent and selective USP30 inhibitor, effective at an IC50 of 120 nM. This compound has been shown to increase protein ubiquitination and accelerate mitophagy, making it suitable for biochemical research applications. It exhibits high selectivity, with less than 30% inhibitory activity against a panel of 22 other USPs at 10 μM.
  • Potent and selective USP30 inhibitor
  • Increases protein ubiquitination
  • Accelerates mitophagy
  • Minimal inhibitory activity against other USPs
  • Stable as powder at -20°C for 3 years
  • Stable in solvent at -80°C for 2 years