Learn More
Medchemexpress LLC HY-112438 5mg Medchemexpress, MF-094 CAS:2241025-68-1 Purity:>98%
SDP
Catalog No. 501932422
Encompass_Preferred
Medchemexpress, HY-112438 5mg MF-094 CAS:2241025-68-1 MF-094 is a potent and selective USP30 inhibitor with an IC 50 of 120 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-193-2422 Supplier Medchemexpress LLC Supplier No. HY1124385MG
Medchemexpress, HY-112438 5mg MF-094 CAS:2241025-68-1 MF-094 is a potent and selective USP30 inhibitor with an IC 50 of 120 nM. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.