Learn More
TARGETMOL CHEMICALS INC MF-438 2MG
SDP
Catalog No. 502877376
Encompass_Preferred
Also available in 1 mL, 1 mg, 5 mg, 10 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. MF-438 is an effective and orally bioavailable stearoyl-CoA desaturase 1 (SCD1) inhibitor (EC50 2.3 nM for rSCD1). Purity 99.66%
Catalog No. 50-287-7376 Supplier TARGETMOL CHEMICALS INC Supplier No. T160682MG
Also available in 1 mL, 1 mg, 5 mg, 10 mg, 25 mg, 50 mg, 100 mg, 500 mg and bulk. Please contact Fisher for quotes. MF-438 is an effective and orally bioavailable stearoyl-CoA desaturase 1 (SCD1) inhibitor (EC50 2.3 nM for rSCD1). Purity 99.66%