Learn More
Medchemexpress LLC HY-15822 5mg Medchemexpress, MF-438 CAS:921605-87-0 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY158225MG
Encompass_Preferred
Medchemexpress, HY-15822 5mg MF-438 CAS:921605-87-0 MF-438 is a potent and orally bioavailable stearoyl-CoA desaturase 1 (SCD1) inhibitor with an EC50 of 2.3 nM for rSCD1. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-203-0686